Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sulf2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003833.2 | NP_001003833.2 | 5 - 6 | 962 | 84 | |
XM_017353509.1 | XP_017208998.1 | 5 - 6 | 972 | 84 | ||
XM_017353510.2 | XP_017208999.1 | 7 - 8 | 1275 | 84 | ||
XM_017353511.2 | XP_017209000.1 | 5 - 6 | 876 | 84 | ||
XM_021469735.1 | XP_021325410.1 | 9 - 10 | 1694 | 84 | ||
XM_021469736.1 | XP_021325411.1 | 4 - 5 | 742 | 84 | ||
XM_021469737.1 | XP_021325412.1 | 4 - 5 | 742 | 84 | ||
XM_021469738.1 | XP_021325413.1 | 4 - 5 | 742 | 84 | ||
XM_021469739.1 | XP_021325414.1 | 4 - 5 | 742 | 84 | ||
GenBank mRNA | AY332607.1 | - | 5 - 6 | 962 | 84 | |
AY332608.1 | - | 4 - 5 | 742 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |