Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wdr55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003871.2 | NP_001003871.1 | 2 - 3 | 188 | 81 | |
GenBank mRNA | AY648800.1 | - | 2 - 3 | 191 | 81 | |
BC095552.1 | - | 2 - 3 | 188 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |