Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004008.1 | NP_001004008.1 | 4 - 5 | 625 | 122 | |
XM_005165300.4 | XP_005165357.1 | 5 - 6 | 740 | 122 | ||
GenBank mRNA | BC080232.1 | - | 4 - 5 | 625 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |