Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crygm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007786.1 | NP_001007787.1 | 2 - 3 | 275 | 77 | |
GenBank mRNA | AY738756.1 | - | 2 - 3 | 275 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |