Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ak5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017540.2 | NP_001017540.1 | 5 - 6 | 976 | 63 | |
GenBank mRNA | BC092184.1 | - | 5 - 6 | 980 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |