Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: epyc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017903.4 | NP_001017903.3 | 6 - 7 | 1669 | 114 | |
NM_001348483.1 | NP_001335412.1 | 4 - 5 | 701 | 114 | ||
XM_005164743.4 | XP_005164800.1 | 7 - 8 | 1884 | 114 | ||
XM_009300279.3 | XP_009298554.1 | 7 - 8 | 1881 | 114 | ||
XM_021475014.1 | XP_021330689.1 | 4 - 5 | 704 | 114 | ||
GenBank mRNA | BC092942.1 | - | 4 - 5 | 517 | 114 | |
BC133975.1 | - | 4 - 5 | 526 | 114 | ||
CU468782.1 | - | 3 - 4 | 488 | 114 | ||
DQ224358.1 | - | 6 - 7 | 1665 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |