Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: thbs4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173226.1 | NP_775333.1 | 12 - 13 | 1665 | 70 | |
GenBank mRNA | AY081009.1 | - | 12 - 13 | 1665 | 70 | |
BC095593.1 | - | 12 - 13 | 1675 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |