Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nitr4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131656.1 | NP_571731.1 | 4 - 5 | 854 | 116 | |
NM_198344.3 | NP_938158.3 | 4 - 5 | 860 | 116 | ||
GenBank mRNA | AF318404.1 | - | 4 - 5 | 854 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Danio rerio Probe spans exons |