Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: her8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199624.3 | NP_955918.3 | 3 - 4 | 329 | 100 | |
GenBank mRNA | AY007990.1 | - | 3 - 4 | 283 | 100 | |
BC068368.1 | - | 3 - 4 | 302 | 100 | ||
EH587922.1 | - | 3 - 4 | 400 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |