Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: foxk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199902.1 | NP_956196.1 | 8 - 9 | 1775 | 109 | |
GenBank mRNA | BC054664.1 | - | 8 - 9 | 1775 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |