Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crispld1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200470.1 | NP_956764.1 | 4 - 5 | 746 | 127 | |
XM_009298567.3 | XP_009296842.1 | 4 - 5 | 769 | 127 | ||
GenBank mRNA | BC055176.1 | - | 4 - 5 | 746 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |