Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: waslb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201059.2 | NP_957353.2 | 7 - 8 | 910 | 85 | |
XM_005164482.4 | XP_005164539.1 | 6 - 7 | 967 | 85 | ||
XM_009299952.3 | XP_009298227.1 | 3 - 4 | 595 | 85 | ||
XM_021474971.1 | XP_021330646.1 | 7 - 8 | 686 | 85 | ||
GenBank mRNA | BC044180.1 | - | 7 - 8 | 960 | 85 | |
BC066424.1 | - | 7 - 8 | 910 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |