Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: veph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212831.1 | NP_997996.1 | 3 - 4 | 555 | 64 | |
GenBank mRNA | AB085188.1 | - | 3 - 4 | 555 | 64 | |
BC134890.1 | - | 3 - 4 | 535 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |