Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myo6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004111.2 | NP_001004111.2 | 14 - 15 | 1559 | 137 | |
XM_005160309.4 | XP_005160366.1 | 15 - 16 | 1705 | 137 | ||
XM_005160311.4 | XP_005160368.1 | 15 - 16 | 1594 | 137 | ||
XM_009294500.3 | XP_009292775.1 | 16 - 17 | 1714 | 137 | ||
XM_009294501.3 | XP_009292776.1 | 16 - 17 | 1714 | 137 | ||
XM_009294502.3 | XP_009292777.1 | 16 - 17 | 1714 | 137 | ||
XM_021468402.1 | XP_021324077.1 | 16 - 17 | 1714 | 137 | ||
XM_021468403.1 | XP_021324078.1 | 14 - 15 | 1553 | 137 | ||
XM_021468404.1 | XP_021324079.1 | 15 - 16 | 1705 | 137 | ||
GenBank mRNA | AY618702.1 | - | 14 - 15 | 1557 | 137 | |
AY691329.1 | - | 15 - 16 | 1600 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Danio rerio Probe spans exons |