Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myo6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004111.2 | NP_001004111.2 | 24 - 25 | 2657 | 86 | |
XM_005160309.4 | XP_005160366.1 | 25 - 26 | 2803 | 86 | ||
XM_005160311.4 | XP_005160368.1 | 25 - 26 | 2692 | 86 | ||
XM_009294500.3 | XP_009292775.1 | 26 - 27 | 2812 | 86 | ||
XM_009294501.3 | XP_009292776.1 | 26 - 27 | 2812 | 86 | ||
XM_009294502.3 | XP_009292777.1 | 26 - 27 | 2812 | 86 | ||
XM_021468402.1 | XP_021324077.1 | 26 - 27 | 2812 | 86 | ||
XM_021468403.1 | XP_021324078.1 | 24 - 25 | 2651 | 86 | ||
XM_021468404.1 | XP_021324079.1 | 25 - 26 | 2803 | 86 | ||
GenBank mRNA | AY618702.1 | - | 24 - 25 | 2655 | 86 | |
AY691329.1 | - | 25 - 26 | 2698 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Danio rerio Probe spans exons |