Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rrm2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007163.2 | NP_001007164.1 | 7 - 8 | 846 | 91 | |
GenBank mRNA | BC108037.1 | - | 7 - 8 | 872 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |