Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chrna5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017885.1 | NP_001017885.1 | 5 - 6 | 480 | 66 | |
XM_021467652.1 | XP_021323327.1 | 4 - 5 | 976 | 66 | ||
GenBank mRNA | BC092962.1 | - | 5 - 6 | 480 | 66 | |
BC164212.1 | - | 4 - 5 | 407 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |