Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020733.2 | NP_001018569.2 | 5 - 6 | 765 | 129 | |
GenBank mRNA | AY584746.1 | - | 5 - 6 | 765 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Danio rerio Probe spans exons |