Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gtpbp1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024811.1 | NP_001019982.1 | 2 - 3 | 311 | 75 | |
XM_005161694.4 | XP_005161751.1 | 3 - 4 | 366 | 75 | ||
XM_009296201.3 | XP_009294476.1 | 3 - 4 | 326 | 75 | ||
GenBank mRNA | BC056806.1 | - | 2 - 3 | 309 | 75 | |
BC096803.1 | - | 2 - 3 | 311 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |