Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpain | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045388.1 | NP_001038853.1 | 6 - 7 | 726 | 128 | |
XM_005171732.4 | XP_005171789.1 | 6 - 7 | 128 | |||
GenBank mRNA | BC122206.1 | - | 6 - 7 | 726 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |