Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: morc3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089366.1 | NP_001082835.1 | 17 - 18 | 2893 | 67 | |
GenBank mRNA | BC135000.1 | - | 17 - 18 | 2893 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |