Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hadhab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089437.1 | NP_001082906.1 | 6 - 7 | 590 | 71 | |
GenBank mRNA | BC093338.1 | - | 6 - 7 | 601 | 71 | |
BC096816.1 | - | 6 - 7 | 615 | 71 | ||
BC107619.1 | - | 6 - 7 | 613 | 71 | ||
BC115107.1 | - | 6 - 7 | 601 | 71 | ||
BC121756.1 | - | 6 - 7 | 601 | 71 | ||
BC131856.1 | - | 6 - 7 | 590 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |