Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbccd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089540.2 | NP_001083009.2 | 6 - 7 | 1896 | 68 | |
GenBank mRNA | BC139633.1 | - | 6 - 7 | 1896 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |