Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100012.2 | NP_001093482.1 | 5 - 6 | 870 | 141 | |
XM_005159014.3 | XP_005159071.1 | 5 - 6 | 1139 | 141 | ||
GenBank mRNA | BC163504.1 | - | 5 - 6 | 1031 | 141 | |
BC163518.1 | - | 5 - 6 | 1031 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |