Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131370.2 | NP_571445.2 | 6 - 7 | 828 | 60 | |
XM_021468355.1 | XP_021324030.1 | 5 - 6 | 753 | 60 | ||
GenBank mRNA | AF143488.1 | - | 6 - 7 | 773 | 60 | |
BC045949.1 | - | 6 - 7 | 828 | 60 | ||
BC165310.1 | - | 6 - 7 | 785 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |