Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scn8aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131628.1 | NP_571703.1 | 4 - 5 | 756 | 125 | |
XM_005162268.4 | XP_005162325.1 | 4 - 5 | 767 | 125 | ||
XM_009296871.3 | XP_009295146.1 | 4 - 5 | 767 | 125 | ||
XM_009296874.3 | XP_009295149.1 | 4 - 5 | 766 | 125 | ||
XM_021469841.1 | XP_021325516.1 | 4 - 5 | 912 | 125 | ||
XM_021469842.1 | XP_021325517.1 | 4 - 5 | 781 | 125 | ||
XM_021469843.1 | XP_021325518.1 | 4 - 5 | 780 | 125 | ||
GenBank mRNA | AF297658.1 | - | 4 - 5 | 756 | 125 | |
BC163773.1 | - | 4 - 5 | 577 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |