Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nme2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199970.2 | 3 - 4 | 314 | 127 | ||
XM_021470954.1 | XP_021326629.1 | 3 - 4 | 396 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Danio rerio Probe spans exons |