Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snapc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200263.1 | NP_956557.1 | 7 - 8 | 790 | 82 | |
GenBank mRNA | BC049022.1 | - | 7 - 8 | 790 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |