Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btr30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200422.1 | NP_956716.1 | 6 - 7 | 1247 | 75 | |
GenBank mRNA | BC054643.1 | - | 6 - 7 | 1247 | 75 |
Gene Symbol: LOC110437745 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021468193.1 | XP_021323868.1 | 5 - 6 | 1166 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |