Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nucb2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201493.1 | NP_958901.1 | 6 - 7 | 778 | 64 | |
GenBank mRNA | BC046077.1 | - | 6 - 7 | 778 | 64 | |
BC065437.1 | - | 6 - 7 | 744 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |