Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ubxn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212607.1 | NP_997772.1 | 7 - 8 | 576 | 95 | |
XM_017358985.2 | XP_017214474.1 | 6 - 7 | 676 | 95 | ||
GenBank mRNA | BC067166.1 | - | 7 - 8 | 576 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |