Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tram2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213071.1 | NP_998236.1 | 7 - 8 | 712 | 143 | |
XM_005173812.3 | XP_005173869.1 | 7 - 8 | 741 | 143 | ||
GenBank mRNA | BC046900.1 | - | 7 - 8 | 712 | 143 | |
BC066535.1 | - | 7 - 8 | 700 | 143 | ||
BC093129.1 | - | 7 - 8 | 712 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |