Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: coq3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002620.2 | NP_001002620.1 | 1 - 2 | 484 | 67 | |
GenBank mRNA | BC075966.1 | - | 1 - 2 | 241 | 67 | |
BC096999.1 | - | 1 - 2 | 484 | 67 | ||
BC164118.1 | - | 1 - 2 | 202 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |