Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rasgef1bb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003479.1 | NP_001003479.1 | 5 - 6 | 906 | 62 | |
GenBank mRNA | BC078340.1 | - | 5 - 6 | 906 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |