Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tm9sf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003550.2 | NP_001003550.1 | 2 - 3 | 370 | 102 | |
XM_021470138.1 | XP_021325813.1 | 2 - 3 | 627 | 102 | ||
GenBank mRNA | BC078291.1 | - | 2 - 3 | 374 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |