Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aqp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003749.4 | NP_001003749.1 | 3 - 4 | 646 | 79 | |
NM_001358313.1 | NP_001345242.1 | 3 - 4 | 646 | 79 | ||
XM_009294620.3 | XP_009292895.1 | 3 - 4 | 804 | 79 | ||
GenBank mRNA | BC078213.1 | - | 3 - 4 | 633 | 79 | |
FJ666327.1 | - | 3 - 4 | 633 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |