Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: drd4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012618.1 | NP_001012636.1 | 2 - 3 | 362 | 79 | |
GenBank mRNA | AY750154.1 | - | 2 - 3 | 362 | 79 | |
BC162965.1 | - | 2 - 3 | 442 | 79 | ||
BC162989.1 | - | 2 - 3 | 442 | 79 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Danio rerio Probe spans exons |