Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnmt3aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001018134.1 | NP_001018144.1 | 5 - 6 | 709 | 78 | |
XM_005160858.4 | XP_005160915.1 | 8 - 9 | 1604 | 78 | ||
GenBank mRNA | AB196919.1 | - | 5 - 6 | 709 | 78 | |
BC162629.1 | - | 4 - 5 | 706 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |