Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rsad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025556.1 | NP_001020727.1 | 5 - 6 | 936 | 114 | |
GenBank mRNA | BC096889.1 | - | 5 - 6 | 936 | 114 | |
BC164179.1 | - | 5 - 6 | 950 | 114 | ||
EF014961.1 | - | 5 - 6 | 931 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |