Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bco2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040312.1 | NP_001035402.1 | 2 - 3 | 276 | 61 | |
GenBank mRNA | BC115259.1 | - | 2 - 3 | 276 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |