Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bco2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040312.1 | NP_001035402.1 | 3 - 4 | 491 | 99 | |
GenBank mRNA | BC115259.1 | - | 3 - 4 | 491 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |