Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bco2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040312.1 | NP_001035402.1 | 9 - 10 | 1312 | 94 | |
GenBank mRNA | BC115259.1 | - | 9 - 10 | 1312 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |