Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srrd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045427.1 | NP_001038892.1 | 4 - 5 | 618 | 65 | |
XM_005172446.4 | XP_005172503.1 | 4 - 5 | 638 | 65 | ||
XM_009303729.3 | XP_009302004.1 | 4 - 5 | 576 | 65 | ||
GenBank mRNA | BC122414.1 | - | 4 - 5 | 618 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |