Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plekhm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001327851.1 | NP_001314780.1 | 11 - 12 | 3215 | 75 | |
NM_001327852.1 | NP_001314781.1 | 10 - 11 | 2576 | 75 | ||
XM_021480128.1 | XP_021335803.1 | 11 - 12 | 3362 | 75 | ||
GenBank mRNA | BC133733.1 | - | 10 - 11 | 2527 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |