Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prdm9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200902.1 | NP_957196.1 | 2 - 3 | 237 | 145 | |
GenBank mRNA | BC064665.1 | - | 2 - 3 | 237 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |