Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prdm9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200902.1 | NP_957196.1 | 5 - 6 | 765 | 78 | |
GenBank mRNA | BC064665.1 | - | 5 - 6 | 765 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |