Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc20a1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212588.1 | NP_997753.1 | 9 - 10 | 1872 | 84 | |
XM_005155625.4 | XP_005155682.1 | 10 - 11 | 2552 | 84 | ||
GenBank mRNA | BC057497.1 | - | 9 - 10 | 1872 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |