Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpl13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212784.1 | NP_997949.1 | 2 - 3 | 200 | 58 | |
GenBank mRNA | BC047855.1 | - | 2 - 3 | 200 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Inventoried Probe spans exons |