Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sirt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002071.1 | NP_001002071.1 | 3 - 4 | 468 | 115 | |
XM_021479673.1 | XP_021335348.1 | 3 - 4 | 485 | 115 | ||
XM_021479674.1 | XP_021335349.1 | 3 - 4 | 330 | 115 | ||
XM_021479677.1 | XP_021335352.1 | 1 - 2 | 210 | 115 | ||
GenBank mRNA | BC071405.1 | - | 3 - 4 | 468 | 115 | |
BC153591.1 | - | 3 - 4 | 435 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |