Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: commd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002421.1 | NP_001002421.1 | 3 - 4 | 347 | 89 | |
GenBank mRNA | BC076149.1 | - | 3 - 4 | 347 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |